Skip to Content
Merck
All Photos(1)

Key Documents

1481000

USP

Oxacillin sodium

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Oxacillin sodium salt monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C19H18N3O5SNa · H2O
CAS Number:
Molecular Weight:
441.43
Beilstein:
4287093
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

oxacillin

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[Na+].[H]O[H].[H][C@]12SC(C)(C)[C@@H](N1C(=O)[C@H]2NC(=O)c3c(C)onc3-c4ccccc4)C([O-])=O

InChI

1S/C19H19N3O5S.Na.H2O/c1-9-11(12(21-27-9)10-7-5-4-6-8-10)15(23)20-13-16(24)22-14(18(25)26)19(2,3)28-17(13)22;;/h4-8,13-14,17H,1-3H3,(H,20,23)(H,25,26);;1H2/q;+1;/p-1/t13-,14+,17-;;/m1../s1

InChI key

ZVIYWUUZWWBNMB-VICXVTCVSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keisuke Nakase et al.
Journal of medical microbiology, 63(Pt 5), 721-728 (2014-02-14)
Propionibacterium acnes and Staphylococcus epidermidis are normal skin inhabitants that are frequently isolated from lesions caused by acne, and these micro-organisms are considered to contribute to the inflammation of acne. In the present study, we examined the antimicrobial susceptibilities and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service