1011743
USP
Adapalene Related Compound D
United States Pharmacopeia (USP) Reference Standard
Synonym(s):
4,4′-Dimethoxy-3,3′-di(adamant-1-yl)biphenyl
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
adapalene
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
O(C)c1c(cc(cc1)c5cc(c(cc5)OC)C76CC8CC(C7)CC(C6)C8)C32CC4CC(C3)CC(C2)C4
InChI
1S/C34H42O2/c1-35-31-5-3-27(13-29(31)33-15-21-7-22(16-33)9-23(8-21)17-33)28-4-6-32(36-2)30(14-28)34-18-24-10-25(19-34)12-26(11-24)20-34/h3-6,13-14,21-26H,7-12,15-20H2,1-2H3
InChI key
SDMSVURKQNRAEU-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Also, for use with USP monograph such as Adapalene
Analysis Note
Other Notes
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service