20081-U
ORBO™-DNPH LpDNPH on Silica Gel, 120 mg
PE separators, O.D. × L 6 mm × 90 mm , w/frangible seal, pk of 10 tubes
Synonym(s):
LpDNPH ORBO™ Tube
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
PE separators
Quality Level
product line
ORBO™
composition
bed wt., 120 mg
packaging
pk of 10 tubes
manufacturer/tradename
ORBO™ DNPH
technique(s)
active air sampling: suitable
O.D. × L
6 mm × 90 mm , w/frangible seal
matrix
DNPH coated silica gel
application(s)
air monitoring
environmental
industrial hygiene
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
The LpDNPH ORBO Tube with frangible seal is designed for determination of low-level carbonyl (e.g formaldehyde) emissions from building materials. The sorbent tube is 1/4 in. O.D. x 90 mm long with frangible design at one end. It contains 120 mg of DNPH coated silica gel and is capped with PTFE caps.
Features and Benefits
- Low background
- Frangible seal design to prevent contamination
Legal Information
ORBO is a trademark of Sigma-Aldrich Co. LLC
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service