Skip to Content
Merck
All Photos(11)

Key Documents

ZRB1534

Sigma-Aldrich

Anti-Collagen X α1 Antibody, clone 1O4 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1O4, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 66.16 kDa
observed mol wt ~60 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1O4 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Collagen X a1. It targets an epitope within 17 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 17 amino acids from the C-terminal region of human Collagen X a1.

Application

Quality Control Testing

Evaluated by Western Blotting in Human skin tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Collagen X a1 in Human skin tissue lysate.

Tested applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Collagen X a1 in Human liver tissue lysate.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Collagen X a1 in human larynx tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound Collagen X a1 with a KD of 1.0 x 10-12 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-Collagen X a1, clone 1O4 ZooMAb®, Cat. No. ZRB1534, is a recombinant Rabbit monoclonal antibody that specifically targets Collagen X a1 and is tested for use in Affinity Binding Assay, Immunohistochemistry (Paraffin), and Western Blotting.

Target description

Collagen alpha-1(X) chain (UniProt: Q03692) is encoded by the COL10A1 gene (Gene ID: 1300) in human. Collagen is the most abundant protein in the body that is predominantly synthesized by fibroblasts. However, epithelial cells may also synthesize small amounts of collagen. The collagen super family includes over 20 different types of collagen with at least 38 distinct polypeptide chains. Collagen X a1 is a homotrimeric protein that is synthesized with a signal peptide (aa 1-18), which is subsequently cleaved off to produce the mature protein that contains two non-helical regions (aa 19-56 and 520-680) and one triple helical region (aa 57-519). Type X a1 collagen is a product of hypertrophic chondrocytes and is localized to mineralization zones of hyaline cartilage. It is a major component of hypertrophic zone and it also influences deposition of other matrix molecules to this region. This provides a proper environment for hematopoiesis, mineralization, and modeling that are essential for endochondral ossification. It is considered as a specific marker of hypertrophic chondrocytes and altered expression of COL10A1 gene is often accompanied by abnormal chondrocyte hypertrophy in many skeletal diseases. Mutations in this gene are reported in Schmid type metaphyseal chondrodysplasia (SMCD), an inherited disorder of the osseous skeleton with short stature, sclerosis of the ribs, and flaring of the metaphyses. Significantly higher expression of COL10A1 gene has also been reported in colorectal cancers. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Huang, H., et al. (2018). Onco Targets Ther. 11; 1571-1581; Gu, J., et al. (2014). Cell Death Dis 5, e1469).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb® formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service