Z720380
Nalgene® centrifuge tubes, Oak Ridge style 3115
nominal capacity 30 mL
Synonym(s):
Nalgene centrifuge tube, plastic centrifuge tube
All Photos(1)
About This Item
UNSPSC Code:
41121703
NACRES:
NB.11
Recommended Products
material
polysulfone
round bottom tube
screw top cap (Linerless)
transparent tube
feature
RCF 50,000 × g
packaging
pack of 10
case of 100
manufacturer/tradename
Nalgene 3115-0030
nominal capacity
30 mL
Related Categories
General description
Strong and tough like polycarbonate but polysulfone much more acid- and base-resistant. Can be used up to 50,000 x g in refrigerated or non-refrigerated centrifuges.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)