Skip to Content
Merck
All Photos(2)

Key Documents

WH0054815M1

Sigma-Aldrich

Monoclonal Anti-GATAD2A antibody produced in mouse

clone 3F3, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-FLJ20085, Anti-GATA zinc finger domain containing 2A, Anti-p66alpha

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3F3, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GATAD2A(54815)

General description

GATAD2A (GATA zinc finger domain containing 2A), a 68kDa protein, is a component of multi-subunit protein Mi-2/NuRD (Nucleosome Remodeling Deacetylase) complex. The Mi-2/NuRD complex is essential for the ATP-dependent chromatin remodeling and histone deacetylase activities. It also performs in the methylation dependent transcriptional repression by guiding repressors to the chromatin. The gene encoding GATAD2A is localized on human chromosome 19p13.11. GATAD2A interacts with the histone tail in vitro and acetylates histone tails.

Immunogen

GATAD2A (NP_060130, 26 a.a. ~ 134 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
QIQKEATAQKPTGSVGSTVTTPPPLVRGTQNIPAGKPSLQTSSARMPGSVIPPPLVRGGQQASSKLGPQASSQVVMPPLVRGAQQIHSIRQHSSTGPPPLLLAPRASVP

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

p66alpha and p66beta of the Mi-2/NuRD complex mediate MBD2 and histone interaction.
Marc Brackertz
Nucleic Acids Research, 34 (2006)
A Drosophila functional evaluation of candidates from human genome-wide association studies of type 2 diabetes and related metabolic traits identifies tissue-specific roles for dHHEX
Jay Pendse
BMC Genomics, 14 (2013)
NURD, a novel complex with both ATP-dependent chromatin-remodeling and histone deacetylase activities
Y Xue
Molecular Cell, 2 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service