SYNP192002
Synthware™ class A graduated pipet
bottle capacity 2 mL, borosilicate glass 3.3
Synonym(s):
Synthware P192002, Pasteur pipette
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass 3.3
packaging
pack of 1 ea
manufacturer/tradename
Kemtech P192002
bottle capacity
2 mL
Looking for similar products? Visit Product Comparison Guide
General description
Pipets, Graduated, Class A, Capacity: 2mL
Application
kemetch; synthware; pipet; graduated; class a
Features and Benefits
Organic Chemistry
Legal Information
Synthware is a trademark of Kemtech America Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service