Skip to Content
Merck
All Photos(1)

Documents

SAB1305978

Sigma-Aldrich

ANTI-IKB ALPHA (IKBA) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

IKBA, MAD3, NF-kappa-B inhibitor alpha, NFKBI, NFKBIA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

35609 Da

species reactivity

human

technique(s)

western blot: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NFKBIA(4792)

Physical form

Supplied in PBS with 0.09% (W/V) sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shirui Tan et al.
Molecular medicine reports, 19(3), 2057-2064 (2019-01-22)
The present study aimed to investigate the anti‑arthritic effects of curculigoside isolated from the rhizome of Curculigo orchioides Gaertn in vivo and in vitro, as well as to determine the potential underlying mechanisms. A rat model of arthritis was induced with type II
Yijia Wang et al.
Gut pathogens, 13(1), 2-2 (2021-01-14)
Inflammatory bowel disease caused by microbial dysbiosis is an important factor contributing to colorectal cancer (CRC) initiation. The 'driver-passenger' model in human gut microbial dysbiosis suggests that 'driver' bacteria may colonize with low relative abundance on tumor site but persistently
Caiguo Zhang et al.
Molecular oncology, 12(4), 476-494 (2018-01-30)
Cullin 4B, a member of the Cullins, which serve as scaffolds to facilitate the assembly of E3 ligase complexes, is aberrantly expressed in many cancers, including osteosarcoma. Recently, we observed that CUL4B forms the CRL4BDCAF11 E3 ligase, which specifically ubiquitinates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service