Skip to Content
Merck
All Photos(1)

Key Documents

PRS2091

Sigma-Aldrich

Anti-CIDE-B (ab3) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Cell death-inducing DNA fragmentation factor, α-subunit-like effector B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 25 kDa

species reactivity

mouse

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Cideb(12684)

Immunogen

a peptide corresponding to amino acids 204-219 of murine CIDE-B .

Linkage

The action of this antibody can be blocked using blocking peptide SBP2091.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service