MWSDS17S
Peptide Molecular Weight Marker
mol wt 2,500-17,000 Da
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
Recommended Products
form
powder
mol wt
glucagon 3,480
myoglobin (1-153) 16,950
myoglobin I (56-131) 8,160
myoglobin I + II (1-131) 14,440
myoglobin I + III (56-153) 10,600
myoglobin II (1-55) 6,210
myoglobin III (132-153) 2,510
2,500-17,000 Da
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
related product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Eduardo L V Silveira et al.
Clinical and vaccine immunology : CVI, 15(8), 1292-1300 (2008-06-27)
Earlier studies have demonstrated in A/Sn mice highly susceptible to Chagas' disease protective immunity against lethal Trypanosoma cruzi infection elicited by vaccination with an open reading frame (ORF) expressed by amastigotes. In our experiments, we used this mouse model to
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service