Key Documents
M9785
Millipore
Ultrafree-MC microcentrifuge filters
working volume 500 μL, Durapore PVDF membrane, pore size 0.65 μm
About This Item
UNSPSC Code:
41103900
NACRES:
NB.24
Pricing and availability is not currently available.
material
Durapore PVDF membrane
polypropylene housing
sterility
non-sterile
manufacturer/tradename
Millipore UFC30DV00
parameter
12,000 × g max. RCF
working volume
500 μL
pore size
0.65 μm
General description
Units consist of microcentrifuge tube with hinged cap and filter insert. The Ultrafree centrifugal filter units are are well suited for the removal of particles & precipitates from aqueous & some solvent-based samples.
Compatibility
Use in any standard fixed-angle microcentrifuge rotor.
Choose from one of the most recent versions:
Certificates of Analysis (COA)