Skip to Content
Merck
All Photos(1)

Key Documents

J1875

Sigma-Aldrich

Jenner′s stain

certified by the Biological Stain Commission, powder

Synonym(s):

Jenner Stain, Eosin−Methylene Blue

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12171500
NACRES:
NA.47
Pricing and availability is not currently available.

Product Name

Jenner′s stain, certified by the Biological Stain Commission, certified by the BSC

grade

certified by the Biological Stain Commission

Agency

certified by the BSC

form

powder

solubility

methanol: 1 mg/mL, blue to very deep blue

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Suitability

Certified for use in staining blood smears.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service