I9409
Monoclonal Anti-Insulin-like Growth Factor-I antibody produced in hamster
clone 126002, purified immunoglobulin, lyophilized powder
Synonym(s):
Anti-IGF-I
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
hamster
Quality Level
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
126002, monoclonal
form
lyophilized powder
species reactivity
mouse
technique(s)
capture ELISA: 2-8 μg/mL
neutralization: suitable
western blot: 1-2 μg/mL
isotype
IgG1
UniProt accession no.
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
mouse ... Igf1(16000)
General description
Insulin-like Growth Factor-I (also known as somatomedinC and somatomedin A) and insulin-like growth factor-II(IGF-II) belong to the family of insulin-like growthfactors, which are structurally homologous to proinsulin.Mature IGF-I and IGF-II are h
Immunogen
purified, E. coli-derived recombinant mouse insulin-like growth factor-I.
Biochem/physiol Actions
Insulin-like growth factor -1 shares structural homology with proinsulin and is mainly produced by liver. IGF-1 acts via specific receptor, IGF-1R, a heterodimeric tyrosine kinase receptor that signals to various second messenger pathways such as MAPK, Raf/Ras and PI3K cascades. The effects of IGF-1R signaling are important at many developmental levels from cells to organ to organism, growth, differentiation, anti-apoptosis, proliferation and transformation.
Physical form
Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
related product
Product No.
Description
Pricing
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
J Dupont et al.
Hormone research, 55 Suppl 2, 22-26 (2001-10-31)
The insulin receptor (IR) and the insulin-like growth factor I receptor (IGF-IR) belong to the same subfamily of receptor tyrosine kinases with two extracellular alpha-subunits and two transmembrane beta-subunits. They share a high similarity of structure and intracellular signalling events.
C J Rosen
Clinical chemistry, 45(8 Pt 2), 1384-1390 (1999-08-03)
The last decade has been characterized by a major investigative thrust into the physiology of two unique but ubiquitous peptides, insulin-like growth factor (IGF)-I and IGF-II. The regulatory systems that control the tissue bioactivity of the IGFs have been delineated
Ettore Capoluongo
The American journal of pathology, 178(1), 26-31 (2011-01-13)
Insulin and insulin-like growth factors (IGFs) are important regulators of energy metabolism and growth. Several findings have outlined an important role played by this family of molecules in both tumor maintenance and development. Despite the established contribution of the IGF
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service