Skip to Content
Merck
All Photos(7)

Key Documents

HPA026749

Sigma-Aldrich

Anti-ASPSCR1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Alveolar soft part sarcoma chromosomal region candidate gene 1 protein, Anti-Alveolar soft part sarcoma locus, Anti-Renal papillary cell carcinoma protein 17, Anti-Tether containing UBX domain for GLUT4, Anti-UBX domain-containing protein 9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

GSSASAGQAAASAPLPLESGELSRGDLSRPEDADTSGPCCEHTQEKQSTRAPAAAPFVPFSGGGQRLGGPPGPTRPLTSSS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ASPSCR1(79058)

General description

Alveolar soft part sarcoma chromosomal region candidate 1 (ASPSCR1) gene is mapped to human chromosome 17q25. ASPSCR1, also known as alveolar soft part sarcoma locus (ASPL) or TUG (tether, containing a UBX domain, for GLUT4) codes for a member of UBX – domain containing proteins. The encoded protein consist of 553 amino acids and it is characterized with a C-terminal ubiquitin regulatory X (UBX)-like domain. ASPSCR1 is ubiquitously expressed in all adult tissues.

Immunogen

Tether containing UBX domain for GLUT4 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Chromosomal alteration at der(17)t(X:17)(p11:q25) leads to the fusion of alveolar soft part sarcoma chromosomal region candidate gene 1 (ASPSCR1) and transcription factor E3 (TFE3) to form the complex ASPSCR1–TFE3 in paraffin-embedded alveolar soft part sarcomas (ASPS).This fusion transcript plays a vital role in differential diagnosis of ASPS. TFE3 forms a complex with ASPL by replacing its N-terminal portion with fused ASPL sequences, but TFE3 retains its DNA-binding domain facilitating transcriptional deregulation in the pathogenesis of ASPS. The protein encoded by ASPSCR1 consists of conserved domains and it might be involved in the ubiquitylation pathway,but specific function of this protein remains unknown. In 3T3-L1 (cell line derived from mouse 3T3 cells) adipocytes, ASPSCR1 controls the trafficking of glucose transporter type 4 (GLUT 4). The encoded protein initiates p97 hexamer disassembly and it is also involved in the reformation of Golgi complex after brefeldin A (BFA) removal.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74813

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Technique for differentiating alveolar soft part sarcoma from other tumors in paraffin-embedded tissue: comparison of immunohistochemistry for TFE3 and CD147 and of reverse transcription polymerase chain reaction for ASPSCR1-TFE3 fusion transcript.
Tsuji K
Human Pathology, 43(3), 356-363 (2012)
The der(17)t(X;17)(p11;q25) of human alveolar soft part sarcoma fuses the TFE3 transcription factor gene to ASPL, a novel gene at 17q25.
Ladanyi M
Oncogene, 20(1), 48-57 (2001)
Detection of the ASPSCR1-TFE3 gene fusion in paraffin-embedded alveolar soft part sarcomas.
Aulmann S
Histopathology, 50(7), 881-886 (2007)
Fusion of a novel gene, RCC17, to the TFE3 gene in t(X;17)(p11.2;q25.3)-bearing papillary renal cell carcinomas.
Heimann P
Cancer Research, 61(10), 4130-4135 (2001)
The ubiquitin regulatory X (UBX) domain-containing protein TUG regulates the p97 ATPase and resides at the endoplasmic reticulum-golgi intermediate compartment.
Orme CM, Bogan JS
The Journal of Biological Chemistry, 287(9), 6679-6692 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service