Skip to Content
Merck
All Photos(4)

Key Documents

HPA007426

Sigma-Aldrich

Anti-TSPAN13 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-NET-6, Anti-TM4SF13, Anti-tetraspanin 13

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

ACLALNQEQQGQLLEVGWNNTASARNDIQRNLNCCGFRSVNPNDTCLASCVKSDHSCSPCAPIIGEYAGEV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TSPAN13(27075)

General description

TSPAN13 (tetraspanin 13) gene encodes a protein belonging to the transmembrane 4 superfamily, also known as the tetraspanin family of proteins. They are integral membrane proteins characterized by the presence of four transmembrane hydrophobic domains. They are 200-300 amino acid long and contain one small extracellular loop of 13-30 amino acids, a short intracellular sequence, and a second, large extracellular loop. This large loop contains a CCG motif (Cys-Cys-Gly) followed by 2-6 cys residues.

Immunogen

tetraspanin 13 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

TSPAN13 (tetraspanin 13) gene is found to be up-regulated in cases of prostate cancer and may play a role in its progression. It is also significantly overexpressed in mucinous, clear cell and endometrioid adenocarcinomas of the ovary, testicular germ cell tumors, bladder carcinoma and B-cell acute lymphoblastic leukaemia. It may serve as a potent breast cancer suppressor gene.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70133

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peng Li et al.
Bosnian journal of basic medical sciences, 19(2), 146-154 (2018-08-17)
Previous studies demonstrated dysregulation of different microRNAs in thyroid cancer. Tetraspanins (TSPANs) are cell surface proteins with critical roles in many cellular processes, and implications in tumor development. Here we investigated the role of miR-369-3p in papillary thyroid cancer (PTC)
Liejun Jiang et al.
Clinical and experimental medicine, 19(3), 377-384 (2019-04-21)
Tetraspanins have been implicated in multiple biological functions including protein networking and cell signaling. NET-6 (TSPAN 13) has been demonstrated to be a tumor suppressor gene in breast cancer, while CD151 is more likely to act as an oncogene. However
Huayi Huang et al.
Journal of cancer research and clinical oncology, 133(10), 761-769 (2007-05-09)
NET-6 is a largely uncharacterized member of the tetraspanin superfamily. We have recently shown that its expression level was lowest in breast carcinomas with aggressive characteristics. We now describe the phenotypic and molecular changes induced in MDA-MB-231 breast carcinoma cells
Antonio Garcia-España et al.
Genomics, 91(4), 326-334 (2008-02-23)
A detailed phylogenetic analysis of tetraspanins from 10 fully sequenced metazoan genomes and several fungal and protist genomes gives insight into their evolutionary origins and organization. Our analysis suggests that the superfamily can be divided into four large families. These
Jose M Arencibia et al.
International journal of oncology, 34(2), 457-463 (2009-01-17)
Prostate cancer is one of the most frequent malignancies in the Western world. The identification of additional molecular markers is needed to refine the diagnosis of prostate cancer and to develop more effective therapies. In order to identify molecular abnormalities

Articles

Learn about glioma markers for high-grade gliomas and low-grade gliomas and find reliable Prestige Antibodies® to target glioma markers.

Learn about glioma markers for high-grade gliomas and low-grade gliomas and find reliable Prestige Antibodies® to target glioma markers.

Learn about glioma markers for high-grade gliomas and low-grade gliomas and find reliable Prestige Antibodies® to target glioma markers.

Learn about glioma markers for high-grade gliomas and low-grade gliomas and find reliable Prestige Antibodies® to target glioma markers.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service