Skip to Content
Merck
All Photos(3)

Key Documents

H4021

Sigma-Aldrich

D-Homoserine

≥98% (TLC)

Synonym(s):

(R)-(+)-2-Amino-4-hydroxybutyric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
119.12
Beilstein:
1721680
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

D-Homoserine,

Assay

≥98% (TLC)

form

powder

color

white

mp

205 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

N[C@H](CCO)C(O)=O

InChI

1S/C4H9NO3/c5-3(1-2-6)4(7)8/h3,6H,1-2,5H2,(H,7,8)/t3-/m1/s1

InChI key

UKAUYVFTDYCKQA-GSVOUGTGSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

D-Homoserine is used for the synthesis of bacterial polysaccharids such as the O-antigen of Acinetobacter lwoffii EK30A. D-Homoserine is used to produce atypical serine protease(s) for mechanism studies.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nikolay P Arbatsky et al.
Organic & biomolecular chemistry, 8(15), 3571-3577 (2010-06-12)
We established a peculiar structure of the O-specific polysaccharide (O-antigen) of a psychrotrophic strain of Acinetobacter lwoffii, EK30A, isolated from a 1.6-1.8 million-year-old Siberian permafrost subsoil sediment sample. The polysaccharide was released by mild acid degradation of the lipopolysaccharide and
Subhash C Annedi et al.
Bioorganic & medicinal chemistry, 14(1), 214-236 (2005-10-04)
The mechanism of proteolysis by serine proteases is a reasonably well-understood process. Typically, a histidine residue acting as a general base deprotonates the catalytic serine residue and the hydrolytic water molecule. We disclose here, the use of an unnatural d-amino
Kimiyasu Isobe et al.
Enzyme research, 2010, 567210-567210 (2010-11-05)
A simple enzymatic method for production of a wide variety of D-amino acids was developed by kinetic resolution of DL-amino acids using L-amino acid oxidase (L-AAO) with broad substrate specificity from Rhodococcus sp. AIU Z-35-1. The optimum pH of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service