EP5490043006
Rotor for Eppendorf® Concentrator Plus
F-45-16-20, non aerosol tight, to hold, 16 x 6.5/10 mL flat-bottom tubes
All Photos(1)
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
Recommended Products
feature
to hold 16 x 20 x 42 - 55 mm
to hold 16 x 6.5/10 mL flat-bottom tubes
fixed angle rotor
non aerosol tight
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5490043006 (EU)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models Concentrator Plus
Legal Information
Eppendorf is a registered trademark of Eppendorf AG