EP0030622103
Eppendorf® Microplate 384/F-PP, 384 wells
white polypropylene, flat bottom clear wells, sterile, pkg of 80 ea
All Photos(1)
About This Item
UNSPSC Code:
41122107
NACRES:
NB.15
Recommended Products
material
flat bottom clear wells
white polypropylene
sterility
sterile; autoclaved
sterile
packaging
pkg of 80 ea
manufacturer/tradename
Eppendorf® 0030622103
technique(s)
PCR: suitable
size
384 wells
Legal Information
Eppendorf is a registered trademark of Eppendorf AG