EP0030000854
Eppendorf® epT.I.P.S.® standard
volume range 0.5-20 μL, pack of 1000 ea (2 bags × 500 tips)
Synonym(s):
pipetting tips
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121600
NACRES:
NB.02
volume:
0.5—20 μL
sterility:
non-sterile
packaging:
pack of 1000 ea (2 bags × 500 tips)
manufacturer/tradename:
Eppendorf® 0030000854
Recommended Products
material
gray
polypropylene
sterility
non-sterile
packaging
pack of 1000 ea (2 bags × 500 tips)
manufacturer/tradename
Eppendorf® 0030000854
availability
available only in (India)
maximum volume
20 μL
volume range
0.5-20 μL
volume
0.5—20 μL
Looking for similar products? Visit Product Comparison Guide
General description
Eppendorf Quality ensures efficient function with tight fitting and precision and precise shape. Low wetting and high chemical and thermal resistance. Good centrifugation stability and excellent material transparency. Color coded pack is dark green containing colorless tips.
Application
Eppendorf® epT.I.P.S.® standard is ideal for dispensing, pipetting, and mixing liquids, filling plates and reaction vessels, filling gels for electrophoresis, and phase extraction and removal of supernatants.
Liquid handling
Features and Benefits
- Original, high-quality Eppendorf pipette tips packaged in resealable bags
- Available in sizes from 10 µL to 10 mL in Eppendorf Quality purity grade epT.I.P.S. Standard 200 µL, 300 µL and 1,000 µL tips are also available color-coded yellow and blue
- epT.I.P.S. Standard pipette tips can be autoclaved at 121 °C in 20 min.
- Fine retracted or extended tips simplify work with small volumes
- Tips are autoclave friendly at 121 °C in 20 min
- High-quality and original pipettes packed in resealable bags
- Fine retracted or extended tips ensure easy pipetting of small sample volumes
- Universal tips that can be used with Eppendorf pipettes as well as pipettes from other manufacturers
- Disposable racks with a slim design, reclosable and easy stackability
- The cone geometry of the tips ensures optimal pipette fit with Eppendorf pipettes
- Fine graduations make visual checks of the pipetting volume even easier
- Long, narrow tips reach the base of narrow and high vessels without touching the top of the vessel edge
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
epT.I.P.S. is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service