Skip to Content
Merck
All Photos(1)

Key Documents

EMU011371

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Map2k2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ATGGCCAACTCGTTTGTAGGGACGCGCTCCTACATGTCCCCAGAGCGGCTGCAGGGCACCCACTACTCTGTGCAGTCGGACATCTGGAGCATGGGGCTGTCGCTGGTGGAGCTGGCCATCGGGAGGTATCCCATTCCCCCACCTGATGCCAAGGAACTAGAGGCCAGCTTTGGCCGGCCTGTGGTGGACGGGGCAGACGGAGAACCCCATAGTGTCTCCCCGAGGCCCAGGCCCCCTGGACGCCCCATCAGTGTAGGTCATGGGATGGACAGCCGACCGGCCATGGCCATCTTTGAGCTGCTGGACTACATAGTGAACGAGCCACCTCCCAAGCTGCCCAGTGGTGTGTTCAGCTCAGACTTCCAGGAGTTTGTGAATAAATGTCTCATTAAGAACCCAGCAGAGCGAGCAGATCTGAAGC

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simona Gargiulo et al.
International journal of molecular sciences, 13(11), 14278-14293 (2012-12-04)
The hypercholesterolemia-atherosclerosis association is now established; hypercholesterolemia may induce vascular-cell activation, subsequently increasing expression of adhesion molecules, cytokines, chemokines, growth factors, and other key inflammatory molecules. Among inflammatory molecules expressed by vascular cells, integrins play a critical role in regulating
Sara Alves et al.
Oncotarget, 6(31), 30787-30802 (2015-09-30)
The recent interest to modulate autophagy in cancer therapy has been hampered by the dual roles of this conserved catabolic process in cancer, highlighting the need for tailored approaches. Since RAS isoforms have been implicated in autophagy regulation and mutation
Caroline N Mills et al.
Molecular cancer, 8, 104-104 (2009-11-19)
Hypoxia inducible factor-1 alpha (HIF-1alpha) protein is rapidly degraded under normoxic conditions. When oxygen tensions fall HIF-1alpha protein stabilizes and transactivates genes involved in adaptation to hypoxic conditions. We have examined the normoxic expression of HIF-1alpha RNA and protein in
Lei-Lei Chen et al.
Autophagy, 13(11), 1969-1980 (2017-09-22)
Recent studies have demonstrated that dysregulation of macroautophagy/autophagy may play a central role in the pathogenesis of neurodegenerative disorders, and the induction of autophagy protects against the toxic insults of aggregate-prone proteins by enhancing their clearance. Thus, autophagy has become
Kazutaka Nanba et al.
Endocrinology, 156(5), 1750-1756 (2015-02-14)
There is considerable evidence supporting the role of calcium signaling in adrenal regulation of both aldosterone synthase (CYP11B2) and aldosterone production. However, there have been no studies that investigated the role played by the Ca(2+)/calmodulin-dependent protein kinase kinase (CaMKK) in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service