CLS8500
Corning® 96 1D/2D bar coded storage tubes
capacity 1.3 mL, without screw caps, bagged
Synonym(s):
sample tube
About This Item
Recommended Products
material
clear tube
polypropylene
round bottom tube
sterility
non-sterile
packaging
pack of 96
case of 960
manufacturer/tradename
Corning
parameter
-180-110 °C temp. range (screw cap)
-80-110 °C temp. range (TPE cap)
L
43.9 mm
capacity
1.3 mL
size
96 wells
tube H
47.5 mm , with TPE cap
52.7 mm , with screw cap
color
none
Looking for similar products? Visit Product Comparison Guide
General description
Features include:
- IDENTIFICATION - Synchronized 2D and linear bar code - unlike any other in the market
- INFORMATION - 14 x 14 dot 2D bar code
- BAR CODE STABILITY - Laser-etched, not an attached label
- COMPOUND COMPATIBILITY - Polypropylene construction throughout
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service