CLS4865
Corning® universal fit pipet tips
1-200 μL universal fit racked pipet tips, yellow, non-sterile, 10 racks/case, 960 tips/case
Synonym(s):
pipet tips, pipette tips
About This Item
RNase free
barrier: no
disposable
endotoxin free
filter: no
graduation
96 Tips/Rack
Recommended Products
material
polypropylene
yellow tip
sterility
non-sterile
feature
DNase free
RNase free
barrier: no
disposable
endotoxin free
filter: no
graduation
96 Tips/Rack
packaging
rack of 96
manufacturer/tradename
Corning 4865
volume range
1-200 μL
volume
1—200 μL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Corning Universal Fit ploypropylene pipet tips are designed to provide a reliable fit with all major brand pipettors and have a bevelled orifice to help ensure fluid delivery. All racked tips are certified RNase/DNase free and nonpyrogenic.
- Reliable fit with all major brand pipettors
- Bevelled orifice to help ensure fluid delivery
- Certified RNase/DNase free and nonpyrogenic
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service