CLS3724
Corning® 1536 well plates
flat bottom black polystyrene wells, non-treated, non-sterile, lid: no
Synonym(s):
1536 multiwell plate, 1536 well microplate, 1536 well microtiter plate, 1536 well plate, HTS plate, TC-treated plate, immunoassay plate, tissue-culture treated plate
About This Item
flat bottom black polystyrene wells
polystyrene plate
material
black plate
flat bottom black polystyrene wells
polystyrene plate
sterility
non-sterile
feature
lid: no
skirt
packaging
pack of 10
case of 50
manufacturer/tradename
Corning 3724
maximum volume
12.5 μL
size
1536 wells
working volume
8 μL
color
black
suitability
suitable for (fluorescent; luminescent assays)
binding type
non-treated surface
Looking for similar products? Visit Product Comparison Guide
General description
- 1536 well polystyrene plates
- Low background fluorescence, minimal light scatter and reduced crosstalk
- Excellent signal dynamic range due to low background fluorescence and enhanced signal intensity
- Opaque walls to prevent well-to-well crosstalk
- Flat well bottoms possess superior flatness -minimizes variability within a plate and between plates
- Round well bottoms for reduced air entrapment and improved CV′s and Z factor
- Raised well bottoms for high sensitivity
- Flood reservior on four sides to reduce instrument contamination
- Suitable for fluorescent and luminescent assays
- Excellent signal dynamic range due to low background fluorescence and enhanced signal intensity
- Superior flatness -minimizes variability within a plate and between plates
- Flat bottom wells are compatible with automated dispensers
- Not treated (or medium binding) polystyrene surface is hydrophobic in nature and binds biomolecules through passive interactions.
- Suitable primarily for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface (binding capacity of approximately 100 to 200ng IgG/cm2)
- Working volume of up to 10 μL
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service