Skip to Content
Merck
All Photos(1)

Key Documents

SHK-1

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 97111106, fish unknown/unspecified, Fibroblast-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

fish unknown/unspecified

packaging

tube of 5 μg 97111106-DNA-5UG
pkg of vial of cells 97111106-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Fibroblast-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Atlantic salmon head kidney

Cell Line Description

The cell line was established from primary cultures of adherent cells from Atlantic salmon head kidney. The cells have been used in the study of the infectious salmon anaemia (ISA) virus. SHK-1 cells are recognised by MAbs specific for Atlantic salmon peripheral blood leucocytes, but not with Mabs specific with polymorphonuclear leucocytes. SHK-1 cells have been shown to phagocytose the fish pathogen Aeromonas salmonicidia to some extent, but no bacterical activity was observed over a period of 72 hours. From these results it has been concluded that SHK-1 is derived from leucocytes with some of the properties of macrophages. Studies have shown that the cells can be passaged more than 60 times.

Application

Propagation of infectious salmon anaemia (ISA) virus
SHK-1 cell line has been used to determine the effects of different nucleotide concentrations on cell viability. It has also been used to study the effects of Silymarin, a compound used in aquaculture.

Culture Medium

Leibovitz L15 + 4mM Glutamine + 40μM 2-Me + 5% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 seeding at 5 x 10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA. Incubate at 20°C. CO2 must not be used. Keep culture dark e.g. by wrapping flask in foil. The cells are sensitive to trypsin which must be removed at each subculture by centrifuging the harvested cells at 100 x g for 5 minutes and resuspending the cell pellet in fresh medium. Cells grow better if conditioned medium is used at each stage supplemented with fresh medium.

Other Notes

Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.
Depositor and originator: Birgit H Dannevig, National Veterinary Institute, Ullevaalsveien 68; Oslo

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service