Skip to Content
Merck
All Photos(1)

Key Documents

CHH-1

92110412, fish cardiovascular , Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

CHH-1, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 92110412, fish cardiovascular, Not specified, 92110412

biological source

fish cardiovascular

packaging

tube of 5 μg 92110412-DNA-5UG
pkg of vial of cells 92110412-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Fish Chum heart

Cell Line Description

Established from normal heart tissue from a juvenile Onchorhynchus keta. Cells are susceptible to a wide range of fish viruses.

Application

Aquatic viral studies

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 using 0.25% trypsin/EDTA; 5% CO2; 24°C. Fish cell lines detach easily during transit if the culture is too young. After resuscitation split cells 1:3 to 1:4 and culture for at least one week with 1-2 medi

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service