Recommended Products
product name
6-23 (clone 6), 87042206
biological source
rat thyroid
growth mode
Adherent
karyotype
Not specified
morphology
Epithelial
products
Neurotensin and calcitonin
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Rat medullary thyroid carcinoma
Cell Line Description
Derived from the non-clonal line rMTC 6-23 originally established from a transplantable WAG/RIJ rat medullary thyroid carcinoma. The cell line synthesises and secretes both neurotensin and calcitonin. Its clonal derivation demonstrates that a population derived from a single type of C-cell has the capability of producing both peptides. Secretion of these peptides has been shown to be stimulated by calcium, KC1, glucagon and catecholamines.
Application
Hormone studies
Culture Medium
DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS). Cells need to be grown on flasks coated with matrigel or any other attachment factor
Subculture Routine
Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C. For cell attachment culture vessels need to be coated with collagen type IV e.g. Sigma product C-5533. Soak in a sol
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service