Skip to Content
Merck
All Photos(1)

Key Documents

SDM103T2

11120712, human lung (effusion), Epitheloid/mesenchymal

Synonym(s):

SDM 103T2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

SDM103T2, from human lung(effusion), 11120712

biological source

human lung (effusion)

growth mode

Adherent

karyotype

Not specified

morphology

Epitheloid/mesenchymal

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

Cell Line Origin

A human mesothelioma cell line derived from a malignant mesothelioma. Asbestos exposure

DNA Profile

STR-PCR Data:
Amelogenin: X,Y
CSF1PO: 11
D13S317: 8
D16S539: 11
D5S818: 11,12
D7S820: 9,10
THO1: 8,9.3
TPOX: 8
vWA: 17

Culture Medium

DMEM:HAMS F12 (1:1) + 2mM Glutamine + 15% Foetal Bovine Serum (FBS)

Subculture Routine

Split subconfluent cultures (70-80%) 1:4 to 1:10 using 0.25% trypsin/EDTA; 5% CO2; 37 °C. Suggested seeding density 1-4x10,000 cells/cm².

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service