LO68 cell line human
10092311, human lung, Epithelial-like: cells are spindle-shaped with few vacuoles
Select a Size
Select a Size
About This Item
Recommended Products
product name
LO68 cell line human, 10092311
biological source
human lung
description
Human malignant mesothelioma
growth mode
Adherent
karyotype
Not specified
morphology
Epithelial-like: cells are spindle-shaped with few vacuoles
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
Cell Line Origin
Application
Culture Medium
Subculture Routine
Other Notes
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service