Skip to Content
Merck
All Photos(1)

Key Documents

C1427

Sigma-Aldrich

Cytochrome b5 human

recombinant, expressed in E. coli (histidine-tagged), ≥90% (SDS-PAGE)

Synonym(s):

Cytochrome b5

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12161501
NACRES:
NA.47

biological source

human

Quality Level

recombinant

expressed in E. coli (histidine-tagged)

Assay

≥90% (SDS-PAGE)

form

solution

mol wt

16 kDa

technique(s)

inhibition assay: suitable

suitability

suitable for molecular biology

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CYB5A(1528)

General description

Cytochrome b5 (CYB5A) gene codes for cytochrome b5 protein. It is located on human chromosome 18. It is co-expressed in the Leydig cells of the testis and theca cells of the ovary.

Biochem/physiol Actions

Cytochrome b5 (CYB5A) mediates NADH-dependent reduction of methemoglobin (erythrocytes), electron transfer to fatty-acid desaturases (mainly liver) and steroid and xenobiotics metabolism by cytochrome P450 enzymes. It is essential for androstenedione synthesis. CYB5A helps in the efficient synthesis of gonadal androgen.
Cytochrome b5 is a membrane-bound heme-containing protein that enhances the catalytic efficiency of some P450 isoforms. Its mechanism of action is unknown.
Membrane-bound heme-containing protein; enhances catalytic efficiency of some P450 isoforms.

Physical form

Solution in 20 mM phosphate buffer, pH 7.25, with 0.1 mM EDTA

Analysis Note

Heme content measured using difference spectroscopy. An extinction coefficient of 100 mM-1 for the absorbance difference between 490 nm and 424 nm is used to calculate the concentration of cytochrome b5.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Melissa R Pitman et al.
Scientific reports, 12(1), 454-454 (2022-01-12)
Sphingosine 1-phosphate (S1P) is a signaling lipid that has broad roles, working either intracellularly through various protein targets, or extracellularly via a family of five G-protein coupled receptors. Agents that selectively and specifically target each of the S1P receptors have
Human adrenal cells that express both beta-hydroxysteroid dehydrogenase type 2 (HSD3B2) and cytochrome b5 (CYB5A) contribute to adrenal androstenedione production
Nakamura Y, et al.
The Journal of Steroid Biochemistry and Molecular Biology, 123(3-5), 122-126 (2011)
CYB5A polymorphism increases androgens and reduces risk of rheumatoid arthritis in women
Stark K, et al.
Arthritis Research & Therapy, 17(1), 56-56 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service