AXYFXF200RS
Pipette tips for Beckman® FX Robotics System
capacity 200 μL, racked, filtered, clear, sterile
Synonym(s):
pipet tips
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
clear
sterility
γ-irradiated
sterile
feature
DNA free
RNase free
barrier: no
endotoxin free
filter
robotic pipet tips
manufacturer/tradename
Axygen Inc
Corning FXF-200-R-S
packaging
pkg of pkg of 5x960 tips/cs
capacity
200 μL
Looking for similar products? Visit Product Comparison Guide
Other Notes
Browse all pipette tips
Legal Information
Beckman is a registered trademark of Beckman Coulter, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service