Skip to Content
Merck
All Photos(1)

Documents

AV45322

Sigma-Aldrich

Anti-OLR1 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-CLEC8A, Anti-LOX1, Anti-Oxidized low density lipoprotein (lectin-like) receptor 1, Anti-SCARE1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

20 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... OLR1(4973)

Immunogen

Synthetic peptide directed towards the N terminal region of human OLR1

Application

Anti-OLR1 antibody produced in rabbit is suitable for western blotting at a concentration of 2.5μg/ml.

Biochem/physiol Actions

Oxidized low density lipoprotein (lectin-like) receptor 1 (OLR1; LOX1) is a receptor for low density lipoprotein (LDL) that belongs to C-type lectin superfamily. It binds, internalizes and degrades oxidized LDL and has a role as scavenger receptor. It may be involved in the regulation of Fas-induced apoptosis. Mutations in OLR1 gene are associated with atherosclerosis and myocardial infarction.

Sequence

Synthetic peptide located within the following region: QANLTHQKKKLEGQISARQQAEEASQESENELKEMIETLARKLNEKSKEQ

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Angela Pirillo et al.
Mediators of inflammation, 2013, 152786-152786 (2013-08-13)
Oxidized low-density lipoprotein (OxLDL) contributes to the atherosclerotic plaque formation and progression by several mechanisms, including the induction of endothelial cell activation and dysfunction, macrophage foam cell formation, and smooth muscle cell migration and proliferation. Vascular wall cells express on
Zufeng Ding et al.
Scientific reports, 3, 2091-2091 (2013-06-29)
Here we demonstrate that "concentration polarization" of ox-LDL enhances LOX-1 expression and ox-LDL uptake. It damages cell surface heparan sulfate proteoglycans (HSPG) and activates LOX-1 dependent autophagy and apoptosis. We also show that ox-LDL concentration polarization occurs on the surface
Miyuki Kumano-Kuramochi et al.
Biochemical and biophysical research communications, 434(3), 594-599 (2013-04-16)
Lectin-like oxidized low-density lipoprotein (LDL) receptor-1 (LOX-1) is an endothelial scavenger receptor that is important for oxidized low-density lipoprotein uptake. LOX-1 functions as an oligomer; however, little is known about the oligomeric complex and ligand processing after recognition by LOX-1.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service