Skip to Content
Merck

Skip To

68360

Fluorescent Red Mega 485 NHS-ester

BioReagent, suitable for fluorescence, ≥90% (coupling to amines)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C29H33N3O9S
CAS Number:
Molecular Weight:
599.65
NACRES:
NA.32
PubChem Substance ID:
UNSPSC Code:
12352108
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioReagent

Quality Segment

assay

≥90% (coupling to amines)

form

powder

fluorescence

λex 482 nm; λem 559 nm in 0.1 M phosphate pH 7.0

suitability

suitable for fluorescence

storage temp.

2-8°C

SMILES string

CCN(CCCCCC(=O)ON1C(=O)CCC1=O)c2ccc3C=C(C(=O)Oc3c2)c4cc[n+](CCCS([O-])(=O)=O)cc4

InChI

1S/C29H33N3O9S/c1-2-31(15-5-3-4-7-28(35)41-32-26(33)10-11-27(32)34)23-9-8-22-19-24(29(36)40-25(22)20-23)21-12-16-30(17-13-21)14-6-18-42(37,38)39/h8-9,12-13,16-17,19-20H,2-7,10-11,14-15,18H2,1H3

InChI key

VKRRBXLGPVGMGA-UHFFFAOYSA-N

Application

Fluorescent Red Mega 485 NHS-ester is an amine conjugating fluorescent biolabel that can be used for covalent coupling to proteins and other biomolecules containing primary amino groups.Fluorescent red Mega 485 and other Mega dyes are specifically designed for multicolor techniques. It is characterized by an extremely large stoke′s shift between excitation and emission maxima. This dye is especially well suited for excitation by argon lasers or other short wavelength excitation light. This means that it can be excited with the same light source or filter arrangement than fluorescein and other widely used dyes as well as other Mega dye. Due to the different emission wavelength several different labels can be detected independently in the same experiment. Due to the small size of the Mega-labels and the direct conversion of excitation light into emission without using a FRET mechanism we foresee a variety of possible applications including DNA sequencing and FISH microscopy.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
93662QBD-AQ-10885LFLGFR100
suitability

suitable for fluorescence

suitability

suitable for fluorescence

suitability

-

suitability

-

fluorescence

λex 482 nm; λem 559 nm in 0.1 M phosphate pH 7.0

fluorescence

λex 672 nm; λem ~735 nm in 0.1 M phosphate pH 7.0

fluorescence

-

fluorescence

-

form

powder

form

powder

form

-

form

pellets

assay

≥90% (coupling to amines)

assay

-

assay

-

assay

-

Quality Level

100

Quality Level

-

Quality Level

100

Quality Level

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-

storage temp.

-


pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters