Skip to Content
Merck
All Photos(1)

Key Documents

43188

Sigma-Aldrich

p-Nitrophenyl-β-D-ribofuranoside

≥98.0% (HPLC), powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13NO7
CAS Number:
Molecular Weight:
271.22
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Product Name

p-Nitrophenyl-β-D-ribofuranoside, ≥98.0% (HPLC)

Quality Level

Assay

≥98.0% (HPLC)

form

powder

impurities

≤0.1% free H-nitrophenol

solubility

methanol: 50 mg/mL, clear, colorless to very faintly yellow

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H](Oc2ccc(cc2)[N+]([O-])=O)[C@H](O)[C@@H]1O

InChI

1S/C11H13NO7/c13-5-8-9(14)10(15)11(19-8)18-7-3-1-6(2-4-7)12(16)17/h1-4,8-11,13-15H,5H2/t8-,9-,10-,11-/m1/s1

InChI key

DUYYBTBDYZXISX-GWOFURMSSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert W P Glowacki et al.
Cell host & microbe, 27(1), 79-92 (2020-01-07)
Efficient nutrient acquisition in the human gut is essential for microbial persistence. Although polysaccharides have been well-studied nutrients for the gut microbiome, other resources such as nucleic acids and nucleosides are less studied. We describe several ribose-utilization systems (RUSs) that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service