PHR2908
Famotidine Related Compound F
certified reference material, pharmaceutical secondary standard
Synonym(s):
3-{[2-(Diaminomethyleneamino)thiazol-4-yl}methylthio]propanoic acid, Famotidine Impurity F (PhEur)
About This Item
Recommended Products
grade
certified reference material
pharmaceutical secondary standard
Quality Level
Agency
traceable to USP 1269265
description
Pharmaceutical Secondary Standard; Certified Reference Material
API family
famotidine
form
solid
packaging
pkg of 50 mg
application(s)
pharmaceutical small molecule
storage temp.
2-8°C
SMILES string
[s]1c(nc(c1)CSCCC(=O)O)NC(=N)N
InChI
1S/C8H12N4O2S2/c9-7(10)12-8-11-5(4-16-8)3-15-2-1-6(13)14/h4H,1-3H2,(H,13,14)(H4,9,10,11,12)
InChI key
JEGZXDCDUSGFSB-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Analysis Note
Other Notes
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service