NIST3124A
Indium (In) standard solution
NIST® SRM® 3124a
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
grade
certified reference material
Quality Level
packaging
pkg of 5 x 10 mL
manufacturer/tradename
NIST®
format
single component solution
Related Categories
General description
This Standard Reference Material (SRM) is intended for use as a primary calibration standard for the quantitative determination of indium. A unit of SRM 3124a consists of five 10 mL sealed borosilicate glass ampoules of solution prepared gravimetrically to contain a known mass fraction of indium. The solution contains nitric acid at a concentration of approximately 1.3 mol/L. For more information, please refer to the SDS and COA.
SRM 3124A_cert
SRM 3124A_SDS
SRM 3124A_cert
SRM 3124A_SDS
Other Notes
Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Indium (In)
See certificate for values and more details at nist.gov/SRM.
Indium (In)
See certificate for values and more details at nist.gov/SRM.
Legal Information
NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B
Supplementary Hazards
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service