Recommended Products
grade
certified reference material
Quality Level
packaging
pkg of 150 g
manufacturer/tradename
NIST®
format
matrix material
Looking for similar products? Visit Product Comparison Guide
General description
This Standard Reference Material (SRM) is low alloy steel in the form of chips sized to pass through sieve openings between 0.50 mm and 1.18 mm (35 mesh to 16 mesh). It is intended primarily for use in evaluation of chemical and instrumental methods of analysis of steel and materials of similar matrix. It should not be used for calibration. A unit of SRM 2166 consists of one bottle containing approximately 150 g of chips. For more information, please refer to the COA and SDS.
SRM 2166_cert
SRM 2166_SDS
SRM 2166_cert
SRM 2166_SDS
Application
- Dental materials and fluoride interaction: Research in Gels explored the influence of fluoride gels on dental materials, including those made from low alloy steel, indicating the material′s suitability for use in environments where chemical interactions are a concern, which is critical for applications in biotechnology and pharmaceutical research (Piszko et al., 2024).
- Biofilm prevention on biometal surfaces: Recent Patents on Biotechnology detailed innovations in biofilm prevention on titanium and stainless steel surfaces, relevant for low alloy steel when considering its applications in biotech equipment that requires stringent hygiene and corrosion resistance (Pires et al., 2020).
Other Notes
Certified for the analytes listed below.
Aluminum (Al), Antimony (Sb), Arsenic (As), Carbon (C), Chromium (Cr), Cobalt (Co), Copper (Cu), Lead (Pb), Manganese (Mn), Molybdenum (Mo), Nickel (Ni), Niobium (Nb), Phosphorus (P), Silicon (Si), Silver (Ag), Sulfur (S), Tin (Sn), Titanium (Ti), Vanadium (V), Sulfur (S)
Reference/Informational value is provided for the following analytes.
Boron (B), Bismuth (Bi), Magnesium (Mg), Selenium (Se), Tantalum (Ta), Tellurium (Te), Zirconium (Zr)
See certificate for values and more details at nist.gov/SRM.
Aluminum (Al), Antimony (Sb), Arsenic (As), Carbon (C), Chromium (Cr), Cobalt (Co), Copper (Cu), Lead (Pb), Manganese (Mn), Molybdenum (Mo), Nickel (Ni), Niobium (Nb), Phosphorus (P), Silicon (Si), Silver (Ag), Sulfur (S), Tin (Sn), Titanium (Ti), Vanadium (V), Sulfur (S)
Reference/Informational value is provided for the following analytes.
Boron (B), Bismuth (Bi), Magnesium (Mg), Selenium (Se), Tantalum (Ta), Tellurium (Te), Zirconium (Zr)
See certificate for values and more details at nist.gov/SRM.
Legal Information
NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service