MBAS1000
Surfactant (MBAS) 1000 mg/L Calibration Standard
certified reference material
Synonym(s):
concentration should be included
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
grade
certified reference material
CofA
current certificate can be downloaded
manufacturer/tradename
RTC MBAS1000-100
concentration
in H2O
technique(s)
UV/Vis spectroscopy: suitable
application(s)
environmental
format
matrix material
storage temp.
2-30°C
Related Categories
Other Notes
Values of analytes vary lot to lot. Some lots may not contain all analytes.
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Surfactant Biodegradation
Swisher RD.
Science, 68-70 (1986)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service