Skip to Content
Merck
All Photos(1)

Key Documents

M0500000

Mepyramine maleate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Pyrilamine maleate salt, Mepyramine maleate salt, N-(4-Methoxyphenyl)methyl-N′,N′-dimethyl-N-(2-pyridinyl)-1,2-ethanediamine maleate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H23N3O · C4H4O4
CAS Number:
Molecular Weight:
401.46
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

pyrilamine, mepyramine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC(=O)\C=C/C(O)=O.COc1ccc(CN(CCN(C)C)c2ccccn2)cc1

InChI

1S/C17H23N3O.C4H4O4/c1-19(2)12-13-20(17-6-4-5-11-18-17)14-15-7-9-16(21-3)10-8-15;5-3(6)1-2-4(7)8/h4-11H,12-14H2,1-3H3;1-2H,(H,5,6)(H,7,8)/b;2-1-

InChI key

JXYWFNAQESKDNC-BTJKTKAUSA-N

Gene Information

human ... HRH1(3269)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Mepyramine maleate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Central hypertensive action of histamine in conscious normotensive cats.
Finch L, et al.
European Journal of Pharmacology, 36(1), 263-266 (1976)
High-performance liquid chromatographic determination of mepyramine maleate, pheniramine maleate and phenylpropanolamine hydrochloride in tablets and drops.
El-Gizawy S M, et al.
Analyst, 112(6), 867-869 (1987)
Lingli Yang et al.
The Journal of investigative dermatology, 134(8), 2105-2113 (2014-03-01)
Histamine is thought to have a critical role in the synthesis of extracellular matrix in skin and may be involved in tissue remodeling of allergic diseases. Recent studies revealed that periostin, a matricelluar protein, contributed to tissue remodeling; however, a
Rhys L Evans et al.
Experimental lung research, 41(4), 189-198 (2015-04-07)
Asthma is associated with reversible airway obstruction, leucocyte infiltration, airways hyperresponsiveness (AHR), and airways remodeling. Fluid accumulation causes pulmonary edema contributing to airways obstruction. We examined the temporal relationship between the late asthmatic response (LAR) following allergen challenge of sensitized
Shigeru Hishinuma et al.
Biochemical pharmacology, 91(2), 231-241 (2014-07-30)
Differential binding sites for first- and second-generation antihistamines were indicated on the basis of the crystal structure of human histamine H1 receptors. In this study, we evaluated differences between the thermodynamic driving forces of first- and second-generation antihistamines for human

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service