Skip to Content
Merck
All Photos(5)

Documents

K4000

Sigma-Aldrich

Kanamycin sulfate from Streptomyces kanamyceticus

Animal Component-free

Synonym(s):

Kanamycin A, Kanamycin sulfate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H36N4O11 · H2O4S
CAS Number:
Molecular Weight:
582.58
EC Number:
MDL number:
UNSPSC Code:
51281654
PubChem Substance ID:
NACRES:
NA.76

biological source

Streptomyces kanamyceticus

Quality Level

form

powder

potency

≥750 μg per mg (dry basis)

color

white to off-white

solubility

H2O: 50 mg/mL, clear, colorless to faintly yellow

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria
mycobacteria
mycoplasma

Mode of action

protein synthesis | interferes

SMILES string

OS(O)(=O)=O.NC[C@H]1O[C@H](O[C@@H]2[C@@H](N)C[C@@H](N)[C@H](O[C@H]3O[C@H](CO)[C@@H](O)[C@H](N)[C@H]3O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C18H36N4O11.H2O4S/c19-2-6-10(25)12(27)13(28)18(30-6)33-16-5(21)1-4(20)15(14(16)29)32-17-11(26)8(22)9(24)7(3-23)31-17;1-5(2,3)4/h4-18,23-29H,1-3,19-22H2;(H2,1,2,3,4)/t4-,5+,6-,7-,8+,9-,10-,11-,12+,13-,14-,15+,16-,17-,18-;/m1./s1

InChI key

OOYGSFOGFJDDHP-KMCOLRRFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Kanamycin Sulfate is a broad spectrum aminoglycoside-antibiotic derived from Streptomyces kanamyceticus.

Application

Kanamycin sulfate is used as an additive in culture media for the isolation of group D streptococci on Kanamycin Esculin Azide Agar and for selection of transformed plant cells containing the neomycin phosphotransferase on a kanamycin-medium. Kanamycin sulfate can also be used as a selection agent for cells transformed with kanamycin B resistance gene. It is recommended for use in cell culture applications at 100 μg/mL. Kanamycin sulfate from Streptomyces kanamyceticus has been used:
  • in the research of bacterial antibiotic resistance
  • in the research on the effect of different antibiotics on bud regeneration

Biochem/physiol Actions

Mode of Action: The product acts by binding to the 70S ribosomal subunit, inhibiting translocation and eliciting miscoding.

Mode of Resistance:Aminoglycoside-modifying enzymes (including acetyltransferase, phosphotransferase, nucleotidyltransferase) can alter this antibiotic, preventing its interaction with ribosomes.

Antimicrobial spectrum: Kanamycin sulfate is effective against gram-negative and gram-postiive bacteria, and mycoplasma.

Features and Benefits

High quality antibiotic suitable for mulitple research applications

Preparation Note

Kanamycin sulfate is soluble in water at 50 mg/mL, yielding a clear solution. It is practically insoluble in alcohol, acetone, chloroform, ether and ethyl acetate. A 1% solution in water has a pH of 6.5 to 8.5. Sterile solutions can be prepared by a sterile filtration, through a .2μm filter.

Solutions are stable at 37°C for approximately 5 days. Aqueous stock solutions can be stored at 2-8°C for long term storage.

Storage and Stability

Tightly closed. Dry. Keep in a well -ventilated place. Keep locked up or in an area accessible

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of gelling agents and antibiotics on adventitious bud regeneration from In vitro leaves of pear
Chevreau et al.
In Vitro Cellular & Developmental Biology. Plant, 33, 173?179-173?179 (1997)
Didrik H Grevskott et al.
Frontiers in microbiology, 8, 24-24 (2017-02-06)
The mechanisms for the development and spread of antibacterial resistance (ABR) in bacteria residing in environmental compartments, including the marine environment, are far from understood. The objective of this study was to examine the ABR rates in Escherichia coli and
Benjamin Bardiaux et al.
Structure (London, England : 1993), 27(7), 1082-1093 (2019-05-06)
Bacterial type 4a pili are dynamic surface filaments that promote bacterial adherence, motility, and macromolecular transport. Their genes are highly conserved among enterobacteria and their expression in enterohemorrhagic Escherichia coli (EHEC) promotes adhesion to intestinal epithelia and pro-inflammatory signaling. To
Marta Lukačišinová et al.
Nature communications, 11(1), 3105-3105 (2020-06-21)
Genetic perturbations that affect bacterial resistance to antibiotics have been characterized genome-wide, but how do such perturbations interact with subsequent evolutionary adaptation to the drug? Here, we show that strong epistasis between resistance mutations and systematically identified genes can be
Fabien J Fuche et al.
Human vaccines & immunotherapeutics, 15(6), 1427-1435 (2018-06-22)
Non-typhoidal Salmonella (NTS) are a leading cause of foodborne infections worldwide, and serogroups B, C1, C2-C3 and D are the most common serogroups associated with human disease. While live vaccine candidates that protect against S. Typhimurium (serogroup B) and S.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service