Skip to Content
Merck
All Photos(1)

Key Documents

80507

Millipore

Bile esculin disks

suitable for microbiology

Synonym(s):

Esculin Bile Disks

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171621
NACRES:
NA.85

product line

BioChemika

form

disc

shelf life

limited shelf life, expiry date on the label

technique(s)

microbe id | specific enzyme detection: suitable

application(s)

clinical testing
food and beverages

microbiology

storage temp.

2-8°C

suitability

Streptococcus spp.

General description

Used for rapid detection of esculin hydrolysis in presence of bile for differentiating group D streptococci from non-group D streptococci. Group D streptococci hydrolyzes the esculin to esculetin and dextrose. Esculetin reacts with an iron salt such as ferric citrate to form a blackish-brown colored complex.

Components

Sterile filter paper discs (50 pcs, diameter 6 mm) impregnated with esculin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

For microbiologists the most fundamental stain was developed in 1884 by the Danish bacteriologist Hans Christian Gram.

For microbiologists the most fundamental stain was developed in 1884 by the Danish bacteriologist Hans Christian Gram.

For microbiologists the most fundamental stain was developed in 1884 by the Danish bacteriologist Hans Christian Gram.

For microbiologists the most fundamental stain was developed in 1884 by the Danish bacteriologist Hans Christian Gram.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service