288500
Silica gel
high-purity grade, pore size 60 Å, 2-22 μm particle size distribution, without binder, suitable for thin layer chromatography (TLC)
About This Item
Recommended Products
grade
high-purity grade
Quality Level
form
powder
manufacturer/tradename
Sigma-Aldrich
technique(s)
thin layer chromatography (TLC): suitable
surface area
~500 m2/g
impurities
≤0.01% Fe3+
particle size distribution
2-22 μm
pore size
60 Å pore size
pH
~6.8 (10% in aq. suspension)
bp
2230 °C
mp
>1600 °C
anion traces
chloride (Cl-): ≤0.01%
SMILES string
O=[Si]=O
InChI
1S/O2Si/c1-3-2
InChI key
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
a. Aquagel - pores are filled with water
b. Xerogel - by the process of evaporation, aqueous phase in the pores are removed
c. Aerogel - solvent removed by supercritical extraction[1]
Application
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service