Skip to Content
Merck
All Photos(2)

Documents

17226

Millipore

Universal Beer Agar

suitable for microbiology, NutriSelect® Plus

Synonym(s):

UB Agar, UBA medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171605
NACRES:
NA.74

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 12 g/L
dextrose, 16.1 g/L
dipotassium phosphate, 0.31 g/L
ferrous sulfate, 0.006 g/L
magnesiumsulfate, 0.12 g/L
manganese sulfate, 0.006 g/L
monopotassium phosphate, 0.31 g/L
peptonized milk, 15 g/L
sodium chloride, 0.006 g/L
tomato juice, 12.2 g/L
yeast extract, 6.1 g/L

manufacturer/tradename

NutriSelect® Plus

packaging

pkg of -500 g

technique(s)

microbiological culture: suitable

final pH

6.3±0.2 (25 °C)

application(s)

bioburden testing
food and beverages

microbiology

suitability

Acinetobacter spp.
Pediococcus spp.
selective for Candida spp.
selective for Escherichia coli
selective for Lactobacillus spp.
selective for Pichia spp.
selective for Proteus spp.
selective for Saccharomyces spp.
selective for Zygosaccharomyces spp.
selective for coliforms

General description

Universal Beer Agar is a base medium supplemented with beer. Its characteristics are closer to natural environmental conditions found in a typical brewery. Thus, it supports the growth of more lactic acid bacteria and yields larger colonies in a shorter time than other brewer’s media. The addition of beer gives the medium some selectivity to the organisms adapted to existing conditions in the brewery. Hop and alcohol inhibit the growth of many other microorganisms, which are not adapted to the beer matrix.

Application

Universal Beer Agar is suitable for culturing spoilage microorganisms of significance in the brewing industry (bacteria like Lactobacillus, Pediococcus, Acetobacter, and yeasts).

Preparation Note

Suspend 62.16 g in 750 ml of distilled water. Heat to boiling to dissolve the medium completely. Add 250 ml beer, without degassing, to the hot medium and mix gently. Dispense as desired and sterilize by autoclaving at 121°C for 10 minutes. If required, add 1 ug/ml (Cat. No. 01810) of Cycloheximide to sterile medium prior to dispensing.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Traditional methods are based morphology, staining methods, enzyme reactions (metabolism) and diverse media.

Cost-effective molecular biology method for lactobacilli detection is presented as an alternative to PCR-based methods.

Cost-effective molecular biology method for lactobacilli detection is presented as an alternative to PCR-based methods.

Cost-effective molecular biology method for lactobacilli detection is presented as an alternative to PCR-based methods.

See All

Protocols

Yeast culture techniques: Model systems for eukaryotic studies with liquid media or agar plate growth.

Yeast culture techniques: Model systems for eukaryotic studies with liquid media or agar plate growth.

Yeast culture techniques: Model systems for eukaryotic studies with liquid media or agar plate growth.

Yeast culture techniques: Model systems for eukaryotic studies with liquid media or agar plate growth.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service