13220
Manganese(II) chloride monohydrate
≥97.0%
Synonym(s):
Manganese dichloride monohydrate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
MnCl2 · H2O
CAS Number:
Molecular Weight:
143.86
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
≥97.0%
form
powder
reaction suitability
core: manganese
reagent type: catalyst
impurities
≤0.01% heavy metals (as Pb)
anion traces
sulfate (SO42-): ≤1000 mg/kg
cation traces
Fe: ≤200 mg/kg
SMILES string
O.Cl[Mn]Cl
InChI
1S/2ClH.Mn.H2O/h2*1H;;1H2/q;;+2;/p-2
InChI key
BXRRQHBNBXJZBQ-UHFFFAOYSA-L
Related Categories
Application
Starting material for preparation of organomanganese chloride reagents; in combination with RLi or RMgX catalyzes reduction of vinyl and aryl halides, stereoselective coupling of vinyl iodides, and alkylation of enol triflates; also used for Mn-Cu-catalyzed conjugate addition of RMgX to enones and for Mn-catalyzed acylation of RMgX by acyl chlorides; in combination with NaBH4 is diastereoselective reducing agent; starting material for the preparation of Mn enolates regio- andstereo-selectively.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - STOT RE 2
Target Organs
Brain
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Cahiez, G.; Alami, M.
Tetrahedron Letters, 30, 3541-3541 (1989)
Fujii, H.; Oshima, K.; Utimoto, K.
Tetrahedron Letters, 32, 6147-6147 (1991)
Cahiez, G.; Laboue, B.
Tetrahedron Letters, 33, 4439-4439 (1992)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service