Skip to Content
Merck
All Photos(1)

Key Documents

MHA000T2B

Millipore

Tryptic Soy Broth- Ready-to-use Bags, Tubes & Ampoules

pack of 50 tests, for use with MF-method, suitable for microbiology

Synonym(s):

CASO, Casein Soybean Digest Broth, Tryptone Soya Broth, SCDB, TSB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106225
eCl@ss:
32160801
NACRES:
NA.74
Pricing and availability is not currently available.

sterility

sterile

Quality Level

form

liquid

shelf life

12 mo.

feature

ready-to-use

packaging

pack of 50 tests

technique(s)

cell counting: suitable (total viable count (TVC))

capacity

2 mL

filling volume

2 mL

color

light amber

input

sample type bottled water
sample type cosmetics
sample type beverage(s)
sample type food(s)
sample type wine
sample type sport drink
sample type cider
sample type beer
sample type soft drink

pH

7.3±0.2 (25.0 °C)

application(s)

clinical
cosmetics
environmental
food and beverages
industrial qc

microbiology

compatibility

for use with MF-method
for use with Microfil®
for use with MicropreSure monitor
for use with MicropreSure®
for use with Milliflex®

shipped in

wet ice

storage temp.

2-8°C

suitability

nonselective for Total count

Looking for similar products? Visit Product Comparison Guide

General description

Tryptic Soy Broth is a general purpose liquid enrichment medium used in qualitative procedures for the sterility test and for the enrichment and cultivation of aerobic and facultatively anaerobic microorganisms that are not excessively fastidious.{18} In clinical microbiology, it may be used for the suspension, enrichment and cultivation of strains isolated on other media. In clinical microbiology, the medium is used in a variety of procedures, e.g., for the preparation of the inoculum and for suspending strains for Kirby-Bauer disc diffusion susceptibility testing and for the microbiological test procedure of culture media. Enzymatic digests of casein and soybean provide amino acids and other complex nitrogenous substances. Glucose (=dextrose) is an energy source. Sodium chloride maintains the osmotic equilibrium. Dibasic potassium phosphate acts as a buffer to control pH. Colonies appear clear to white colonies; some may produce pigment.

Application

Tryptone Soy Broth is used as a non-selective medium used to detect total heterotrophic microorganisms in water, dairy products and other liquids like Beer, Bottled Water, Cider, Soft Drink, Sport Drink and Wine. The media finds its application in Clinical, Cosmetics, Environmental Analysis, Food and Beverage and Industrial QC.

Packaging

2 mL plastic ampoules

Specifications

Incubation Temperature: 25–35 °C
Incubation Time: 48–72 hrs

Culture Medium

Trypticase Soy

Analysis Note

Clear to white colonies; some may produce pigment.
QC organisms
S. epidermidis (ATCC 14990)

C. albicans (ATCC 10231)

E. coli (ATCC 25922)

B. subtilis (ATCC 6633)

A.brasilliensisformerly A. niger (ATCC 16404 / WDCM 00053)

P. aeruginosa (ATCC 27853)
Target Microorganisms: Total Count

Legal Information

MICROFIL is a registered trademark of Merck KGaA, Darmstadt, Germany
MICROPRESURE is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diagnostic microbiology
Forbe BA et al.
Bailey and Scott's Diagnostic Microbiology, 11 (1988)
Media for Isolation-Identification-Cultivation-Maintenance of Medical Bacteria
MacFaddin JF
Williams and Wilkins (1985)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service