Skip to Content
Merck
All Photos(3)

Documents

MABF2053

Sigma-Aldrich

Anti-CD117 (c-Kit) Antibody, clone 2C11

clone 2C11, from mouse

Synonym(s):

Mast/stem cell growth factor receptor Kit, EC: 2.7.10.1, SCFR, Piebald trait protein, PBT, Proto-oncogene c-Kit, Tyrosine-protein kinase Kit, p145 c-kit, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

2C11, monoclonal

species reactivity

mouse, human

packaging

antibody small pack of 25 μg

technique(s)

flow cytometry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... KIT(3815)

General description

Mast/stem cell growth factor receptor Kit (UniProt: P10721; also known as EC: 2.7.10.1, SCFR, Piebald trait protein, PBT, Proto-oncogene c-Kit, Tyrosine-protein kinase Kit, p145 c-kit, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog) is encoded by the KIT (also known as SCFR) gene (Gene ID: 3815) in human. C-Kit is a single-pass type I membrane protein with tyrosine protein kinase activity and serves as a cell-surface receptor for the cytokine KITLG/SCF and is shown to play a significant role in the regulation of cell survival, cell proliferation, stem cell maintenance, gametogenesis and other functions. c-Kit exists as a monomer in the absence of bound KITLG/SCF, however, it homodimerizes in the presence of bound KITLG/SCF and forms a heterotetramer with two KITLG/SCF molecules. c-Kit can be autophosphorylated on tyrosine residues and KITLG/SCF binding enhances its autophosphorylation. Its kinase activity is down-regulated by phosphorylation on serine residues by PKC family members. Phosphorylation at Tyrosine 568 is essential for its interaction with PTPN11/SHP-2, CRK (isoform Crk-II) and members of the SRC tyrosine-protein kinase family. c-Kit can be ubiquitinated by SOCS6 and it is rapidly ubiquitinated after autophosphorylation induced by KITLG/SCF binding, leading to its internalization and degradation. c-Kit is synthesized with a signal peptide (aa 1-25), which is subsequently cleaved off to produce the mature form with an extracellular domain (aa 26-524), a short transmembrane domain (aa 525-545), and a cytoplasmic domain (aa 546-976. Its two ATP-binding sites are localized to amino acids 596-603 and 671-677. Lack of c-Kit expression has been reported in multiple cancer types and c-Kit expressing cells display a regression of typical morphological features of malignancy.

Specificity

Clone 2C11 specifically detects CD117 (c-Kit) in human and murine cells. It targets an epitope with in the extracellular domain.

Immunogen

GST-tagged recombinant fragment corresponding to 195 amino acids from the extracellular domain of Human C-kit.

Application

Flow Cytometry Analysis: A representative lot detected CD117 (c-Kit) in P815 cells (Courtesy of the Monoclonal Antibody Unit at the Institute of Molecular and Cell Biology, Agency for Science, Technology and Research (A*STAR), Singapore).

Western Blotting Analysis: A representative lot detected CD117 (c-Kit) in DLD-1 cell lysate (Courtesy of the Monoclonal Antibody Unit at the Institute of Molecular and Cell Biology, Agency for Science, Technology and Research (A*STAR), Singapore).
This mouse monoclonal Anti-CD117 (c-Kit), clone 2C11 Antibody, Cat. No. MABF2053 is tested for use in Western Blotting and Flow Cytometry, for the detection of C-kit.

Quality

Evaluated by Western Blotting in DLD-1 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected CD117 (c-Kit) in DLD-1 cell lysate.

Target description

~160 kDa observed; 109.87 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service