Skip to Content
Merck
All Photos(3)

Documents

MABC1023

Sigma-Aldrich

Anti-EGR1 Antibody, clone 16A10.1

clone 16A10.1, from mouse

Synonym(s):

Early growth response protein 1, AT225, EGR-1, Nerve growth factor-induced protein A, NGFI-A, Transcription factor ETR103, Transcription factor Zif268, Zinc finger protein 225, Zinc finger protein Krox-24

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

16A10.1, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... EGR1(1958)

General description

Early growth response protein 1 (UniProt P18146; also known as AT225, EGR-1, Nerve growth factor-induced protein A, NGFI-A, Transcription factor ETR103, Transcription factor Zif268, Zinc finger protein 225, Zinc finger protein Krox-24) is encoded by the EGR1 (also known as KROX24, ZNF225) gene (Gene ID 1958) in human. EGR-1 is the founding member of the early growth response (EGR) family of Cys2-His2-type zinc finger transcription factors that share a high homology in their DNA-binding region. EGR-1 is ubiquitously expressed in many cell types and binds to a consensus DNA motif (GCGGTGGGCG) in target genes, including c-Myc, cyclins D2 & G2, p19, TNF, IL-2, CD44, and ICAM-1. EGR1 is known as an immediate-early response gene due to the rapid kinetics of its induction by various signals, including growth factors, cytokines, and stress. Human EGR-1 is a 543-amino acid protein with three C2H2-type Zinc fingers (a.a. 338-362, 368-390, 396-418).

Immunogen

Epitope: Internal.
GST-tagged recombinant protein corresponding to an internal fragment of human EGR1.

Application

Anti-EGR1 Antibody, clone 16A10.1 is an antibody against EGR1 for use in Western Blotting, Immunocytochemistry.
Immunocytochemistry Analysis: An 1:250 dilution of this antibody from a representative lot detected EGF-induced upregulation of nuclear EGR1 immunoreactivity in serum-starved HeLa and NIH/3T3 cells.
Research Category
Apoptosis & Cancer
Research Sub Category
Transcription Factors

Quality

Evaluated by Western Blotting in HUVEC lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected VEGF-induced EGR1 expression in serum-starved HUVECs.

Target description

~80 kDa observed. 57.51 kDa calculated. The observed target band size is consistent with the reported apparent molecular weight (Krones-Herzig, A., et al. (2005). Cancer Res. 65(12):5133-5143; Josefsen, K., et al. (1999). Diabetologia. 42(2):195-203).

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service