Skip to Content
Merck
All Photos(1)

Key Documents

MAB349

Sigma-Aldrich

Anti-Beta (β)-Amyloid antibody

CHEMICON®, mouse monoclonal, 1.D5

Synonym(s):

Alz-90

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Product Name

Anti-Amyloid Precursor Protein Antibody, A4(695), clone 1.D5, clone 1.D5, Chemicon®, from mouse

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1.D5, monoclonal

species reactivity

human, rat

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... APP(351)

Specificity

Reacts with the natural amyloid precursor protein A4(695).

Immunogen

Epitope: A4(695)
Synthetic peptide Alz 90 [amino acids 511-608 of amyloid precursor protein pre A4(695)].

Application

Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases
This Anti-Amyloid Precursor Protein Antibody, A4(695), clone 1.D5 is validated for use in WB for the detection of Amyloid Precursor Protein.
Western blot: 10-20 μg/mL

Optimal working dilutions must be determined by the end user.

Westen Blot Protocol for MAB349

1) After blotting the SDS-PAGE gel onto nitrocellulose, incubate at room temperature for one hour in 1% PBS in 0.1 mol/L PBS, pH 7.2, to block non-specific background.

2) Incubate the Western blot with MAB349 (10-20 μg/mL) at +4°C overnight shaking well.

3) Wash the Western blots 2 times in PBS-Tween 0.05% for ten minutes. Wash one time in PBS.

4) Incubate the blots with anti-Mouse IgG (Chemicon AP124) for one hour at room temperature.

5) Repeat step #3.

6) Incubate in peroxidase-anti-peroxidase [PAP] (Chemicon PAP14) according to manufacturers directions.

7) Repeat step #3.

8) Incubate in 4-chloro-1-napthol for twenty minutes at room temperature.

9) Repeat step #3.

Physical form

Format: Purified
Liquid in 0.02M Phosphate buffer, 0.25M NaCl, pH 7.6 with 0.1% sodium azide.

Storage and Stability

Maintain lyophilized material at +2-8°C for up to 12 months. After reconstitution maintain frozen at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christine Villegas et al.
Human molecular genetics, 23(6), 1631-1643 (2013-11-10)
The amyloid-β precursor protein (APP), a type I transmembrane protein genetically associated with Alzheimer's disease, has a complex biology that includes proteolytic processing into potentially toxic fragments, extensive trafficking and multiple, yet poorly-defined functions. We recently proposed that a significant
Virgil Muresan et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 29(11), 3565-3578 (2009-03-20)
The amyloid-beta (Abeta) precursor protein (APP), a transmembrane protein that undergoes proteolytic cleavage into defined fragments, has been implicated in axonal transport. The proposed role of APP as a vesicle receptor for the microtubule motor kinesin-1 has relevance for the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service