CPS1
mPAGE® Casting Plate Rack
for use with mPAGE® Mini Spacer Plates and Mini Short Plates
Synonym(s):
drying rack
About This Item
Recommended Products
packaging
pkg of 1 pack
manufacturer/tradename
Millipore
technique(s)
SDS-PAGE: suitable
electrophoresis: suitable
General description
The mPAGE® Gel Caster offers a leak-free solution for handcasting up to 2 mini protein gels. Gels cast
are 8 cm x 10 cm with a gel thickness of 1.0 mm, 0.75 mm, or 1.5 mm, along
with 10 or 15-well options for sample loading. The mPAGE® Gel Caster has been
designed to simplify hand casting gels and to eliminate leakage during the casting
process. The gel caster is available as a standalone device (GCR2) or as a gel casting device kit for casting mini gels gels with 0.75 mm, 1.0 mm and 1.5 mm thickness (MGCK-75M, MGCK-10M, MGCK-15M).
Compatibility
Legal Information
related product
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protocols
Introduction to PAGE. Learn about SDS-PAGE background and protocol for the separation of proteins based on size in a poly-acrylamide gel.
Introduction to PAGE. Learn about SDS-PAGE background and protocol for the separation of proteins based on size in a poly-acrylamide gel.
Introduction to PAGE. Learn about SDS-PAGE background and protocol for the separation of proteins based on size in a poly-acrylamide gel.
Introduction to PAGE. Learn about SDS-PAGE background and protocol for the separation of proteins based on size in a poly-acrylamide gel.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service