Skip to Content
Merck
All Photos(2)

Key Documents

8.21125

Sigma-Aldrich

Toluene-4-sulfonic acid sodium salt

for synthesis

Synonym(s):

Toluene-4-sulfonic acid sodium salt, 4-Toluenesulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(CH3)C6H4SO3Na
CAS Number:
Molecular Weight:
194.18
MDL number:
UNSPSC Code:
12352107
EC Index Number:
211-522-5
NACRES:
NA.22

vapor pressure

<1 hPa ( 20 °C)

Quality Level

form

powder

mp

>300 °C

solubility

soluble 818 g/L

density

1.55 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[Na+].[S](=O)(=O)([O-])c1ccc(cc1)C

InChI

1S/C7H8O3S.Na/c1-6-2-4-7(5-3-6)11(8,9)10;/h2-5H,1H3,(H,8,9,10);/q;+1/p-1

InChI key

KVCGISUBCHHTDD-UHFFFAOYSA-M

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

932.0 °F - closed cup

Flash Point(C)

500 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service