Skip to Content
Merck
All Photos(1)

Documents

8.11689

Sigma-Aldrich

Kryptofix® 5

for synthesis

Synonym(s):

Kryptofix® 5, 1,13-Bis(8-quinolyl)-1,4,7,10,13-pentaoxatridecane, 8,8-(1,4,7,10,13-Pentaoxatridecylene)-diquinoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H28N2O5
CAS Number:
Molecular Weight:
448.51
UNSPSC Code:
12352112
EC Index Number:
260-674-9
NACRES:
NA.22

Quality Level

form

powder

mp

72-76 °C

storage temp.

2-30°C

InChI

1S/C26H28N2O5/c1-5-21-7-3-11-27-25(21)23(9-1)32-19-17-30-15-13-29-14-16-31-18-20-33-24-10-2-6-22-8-4-12-28-26(22)24/h1-12H,13-20H2

InChI key

GHYUCQLUMXYVRT-UHFFFAOYSA-N

Application

Kryptofix® 5 for synthesis. CAS 57310-75-5, molar mass 448.52 g/mol.

Analysis Note

Assay (HClO₄): ≥ 99.0 %
Melting range (lower value): ≥ 72 °C
Melting range (upper value): ≤ 76 °C
Identity (IR): passes test

Legal Information

Kryptofix is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service